Learn More
Alfa Aesar™ 1-tert-Butyl-4-nitrobenzene, 98%
Brand: Alfa Aesar™ L06067.18
Additional Details : CAS Number : 3282-56-2 Weight : 0.05000kg
Quantity | 50g |
---|
Chemical Identifiers
3282-56-2 | |
179.219 | |
XSCPVQNNFLHGHY-UHFFFAOYSA-N | |
76773 | |
CC(C)(C)C1=CC=C(C=C1)[N+](=O)[O-] |
C10H13NO2 | |
MFCD00041253 | |
1-tert-butyl-4-nitrobenzene, 4-t-butylnitrobenzene, 4-tert-butylnitrobenzene, p-t-butylnitrobenzene, p-nitro-t-butylbenzene, 4-t-butyl nitrobenzene, benzene, 1-1,1-dimethylethyl-4-nitro, tert-butyl-4-nitrobenzene, acmc-209hw6, benzene,1-dimethylethyl-4-nitro | |
1-tert-butyl-4-nitrobenzene |
Specifications
1-tert-Butyl-4-nitrobenzene | |
1.534 | |
50g | |
C10H13NO2 | |
2047337 | |
XSCPVQNNFLHGHY-UHFFFAOYSA-N | |
1-tert-butyl-4-nitrobenzene | |
76773 | |
98% |
1.0586 | |
265°C to 267°C | |
3282-56-2 | |
MFCD00041253 | |
1-tert-butyl-4-nitrobenzene, 4-t-butylnitrobenzene, 4-tert-butylnitrobenzene, p-t-butylnitrobenzene, p-nitro-t-butylbenzene, 4-t-butyl nitrobenzene, benzene, 1-1,1-dimethylethyl-4-nitro, tert-butyl-4-nitrobenzene, acmc-209hw6, benzene,1-dimethylethyl-4-nitro | |
CC(C)(C)C1=CC=C(C=C1)[N+](=O)[O-] | |
179.219 | |
179.22 |