missing translation for 'onlineSavingsMsg'
Learn More

1-Octen-3-ol, 98%, Thermo Scientific Chemicals

Catalog Number p-3758585
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
3391-86-4
C8H16O
128.22
MFCD00004589
VSMOENVRRABVKN-UHFFFAOYSA-N
1-octen-3-ol, vinyl amyl carbinol, 1-vinylhexanol, 3-hydroxy-1-octene, amyl vinyl carbinol, mushroom alcohol, matsutake alcohol, vinyl hexanol, pentyl vinyl carbinol, matsuica alcohol
18827
CHEBI:34118
oct-1-en-3-ol
CCCCCC(C=C)O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
3391-86-4
C8H16O
128.22
MFCD00004589
VSMOENVRRABVKN-UHFFFAOYSA-N
1-octen-3-ol, vinyl amyl carbinol, 1-vinylhexanol, 3-hydroxy-1-octene, amyl vinyl carbinol, mushroom alcohol, matsutake alcohol, vinyl hexanol, pentyl vinyl carbinol, matsuica alcohol
18827
CHEBI:34118
oct-1-en-3-ol
CCCCCC(C=C)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8370g/mL
Boiling Point 174.0°C
Flash Point 68°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)4CH(OH)CH=CH2
Refractive Index 1.4340 to 1.4420
Quantity 5 g
Specific Gravity 0.837
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in ethanol
Formula Weight 128.22
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Octen-3-ol
Show More Show Less
compliance-icons
Product Identifier
  • 1-Octen-3-ol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'