Learn More
1-Naphthalenemethylamine, 98+%, Thermo Scientific Chemicals
Catalog Number p-7043935
Click to view available options
Quantity:
1 g
5 g
25 g
118-31-0
C11H12N
158.22
MFCD00004048
NVSYANRBXPURRQ-UHFFFAOYSA-O
1-naphthalenemethylamine, 1-naphthalenemethanamine, 1-naphthylmethylamine, 1-aminomethyl naphthalene, 1-naphthalenemethyl amine, c-naphthalen-1-yl-methylamine, 1-naphthylmethanamine, 1-aminomethylnaphthalene, naphthylmethylamine, 1-naphthylmethyl amine hydrochloride
8355
naphthalen-1-ylmethanamine
[NH3+]CC1=C2C=CC=CC2=CC=C1
118-31-0
C11H12N
158.22
MFCD00004048
NVSYANRBXPURRQ-UHFFFAOYSA-O
1-naphthalenemethylamine, 1-naphthalenemethanamine, 1-naphthylmethylamine, 1-aminomethyl naphthalene, 1-naphthalenemethyl amine, c-naphthalen-1-yl-methylamine, 1-naphthylmethanamine, 1-aminomethylnaphthalene, naphthylmethylamine, 1-naphthylmethyl amine hydrochloride
8355
naphthalen-1-ylmethanamine
[NH3+]CC1=C2C=CC=CC2=CC=C1
1-Naphthalenemethylamine is used to increase the induced circular dichroism (ICD) magnitude exhibited by poly[(4-carboxyphenyl)acetylene]. Further, it reacts with monomethoxypoly(ethylene glycol) succinimido carbonate to prepare carbamate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Naphthalenemethylamine is used to increase the induced circular dichroism (ICD) magnitude exhibited by poly[(4-carboxyphenyl)acetylene]. Further, it reacts with monomethoxypoly(ethylene glycol) succinimido carbonate to prepare carbamate.
Solubility
Miscible with ethanol, ether and carbon disulfide.
Notes
Air sensitive. Incompatible with acids, acid chlorides, acid anhydrides, oxidizing agents and chloroformates.
TRUSTED_SUSTAINABILITY
Density 1.075
Boiling Point 289°C to 292°C
Flash Point >110°C (230°F)
Refractive Index 1.643
Quantity 1 g
UN Number UN2735
Beilstein 2206459
Sensitivity Air sensitive; May form a precipitate
Solubility Information Miscible with ethanol,ether and carbon disulfide.
Formula Weight 157.22
Percent Purity ≥98%
Chemical Name or Material 1-Naphthalenemethylamine