missing translation for 'onlineSavingsMsg'
Learn More

1-N-BOC-Aminocyclopentanecarboxylic acid, 98%, Thermo Scientific™

Catalog Number p-2689583
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
35264-09-6
C11H19NO4
229.28
MFCD01076126
YBZCSKVLXBOFSL-UHFFFAOYSA-N
boc-cycloleucine, 1-boc-amino cyclopentanecarboxylic acid, 1-tert-butoxycarbonylamino cyclopentanecarboxylic acid, 1-n-boc-aminocyclopentanecarboxylic acid, boc-1-aminocyclopentane-1-carboxylic acid, n-boc-cycloleucine, 1-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1-tert-butoxycarbonylamino-cyclopentanecarboxylic acid, 1-tert-butoxy carbonyl amino cyclopentane-1-carboxylic acid
2734645
1-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid
CC(C)(C)OC(=O)NC1(CCCC1)C(=O)O
This item is not returnable. View return policy
35264-09-6
C11H19NO4
229.28
MFCD01076126
YBZCSKVLXBOFSL-UHFFFAOYSA-N
boc-cycloleucine, 1-boc-amino cyclopentanecarboxylic acid, 1-tert-butoxycarbonylamino cyclopentanecarboxylic acid, 1-n-boc-aminocyclopentanecarboxylic acid, boc-1-aminocyclopentane-1-carboxylic acid, n-boc-cycloleucine, 1-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1-tert-butoxycarbonylamino-cyclopentanecarboxylic acid, 1-tert-butoxy carbonyl amino cyclopentane-1-carboxylic acid
2734645
1-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid
CC(C)(C)OC(=O)NC1(CCCC1)C(=O)O
Melting Point 130.0°C to 135.0°C
CAS Min % 97.5
CAS Max % 100.0
Color White
Assay Percent Range 98%
Infrared Spectrum Authentic
Packaging Glass Bottle
Solubility Information Solubility in water: soluble in hot water. Other solubilities: soluble in methanol,insoluble in toluene
Formula Weight 229.28
Percent Purity 98%
Quantity 1 g
Physical Form Crystalline Powder
Chemical Name or Material 1-N-BOC-Aminocyclopentanecarboxylic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'