missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Methylpiperidine-2-methanol, 98%

Brand:  Alfa Aesar™ B22187.14

Additional Details : CAS Number : 20845-34-5 Weight : 0.02500kg Transport : UN number : 2735 Chem class : 8 Pack group : II

Quantity 25g

 View more versions of this product

Product Code. 11389759

  • 608.00 NOK / 25g


Estimated Shipment
28-01-2021
Add to basket

Description

Description

Specifications

Chemical Identifiers

20845-34-5
129.203
HXXJMMLIEYAFOZ-UHFFFAOYSA-N
89394
CN1CCCCC1CO
C7H15NO
MFCD00006494
1-methyl-2-piperidinemethanol, 1-methylpiperidin-2-yl methanol, 1-methylpiperidine-2-methanol, 2-piperidinemethanol, 1-methyl, 1-methyl-2-piperidyl methanol, 1-methyl-2-hydroxymethylpiperidine, 2-hydroxymethyl-1-methylpiperidine, 1-methyl-2-piperidyl methan-1-ol, pubchem8873, 1-methyl-2piperidinemethanol
(1-methylpiperidin-2-yl)methanol

Specifications

1-Methylpiperidine-2-methanol
81°C (177°F)
79°C to 80°C (7mmHg)
20845-34-5
MFCD00006494
1-methyl-2-piperidinemethanol, 1-methylpiperidin-2-yl methanol, 1-methylpiperidine-2-methanol, 2-piperidinemethanol, 1-methyl, 1-methyl-2-piperidyl methanol, 1-methyl-2-hydroxymethylpiperidine, 2-hydroxymethyl-1-methylpiperidine, 1-methyl-2-piperidyl methan-1-ol, pubchem8873, 1-methyl-2piperidinemethanol
CN1CCCCC1CO
129.203
129.2
0.984
1.483
25g
C7H15NO
103612
HXXJMMLIEYAFOZ-UHFFFAOYSA-N
(1-methylpiperidin-2-yl)methanol
89394
98%
Videos
SDS Documents