missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methylnaphthalene, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
1-Methylnaphthalene is used as an important intermediate in organic synthesis. It is used as a starting material of surfactant, water reducer, dispersant and drugs. Further, it is employed in the preparation of 1-methylnaphthalene-d10 using deuterium oxide, sodium deuteroxide
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Methylnaphthalene is used as an important intermediate in organic synthesis. It is used as a starting material of surfactant, water reducer, dispersant and drugs. Further, it is employed in the preparation of 1-methylnaphthalene-d10 using deuterium oxide, sodium deuteroxide
Solubility
Miscible with water, alcohol, ethanol, ether and benzene.
Notes
Store in a cool place. Incompatible with strong oxidizing agents, peroxides and oxygen.
1-Methylnaphthalene is used as an important intermediate in organic synthesis. It is used as a starting material of surfactant, water reducer, dispersant and drugs. Further, it is employed in the preparation of 1-methylnaphthalene-d10 using deuterium oxide, sodium deuteroxide
Solubility
Miscible with water, alcohol, ethanol, ether and benzene.
Notes
Store in a cool place. Incompatible with strong oxidizing agents, peroxides and oxygen.
Specifications
Melting Point | -22°C |
Boiling Point | 240°C to 243°C |
Flash Point | 82°C (179°F) |
Quantity | 25 g |
UN Number | UN3082 |
Beilstein | 506793 |
Formula Weight | 142.2 |
Percent Purity | 96% |
Chemical Name or Material | 1-Methylnaphthalene |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'