missing translation for 'onlineSavingsMsg'
Learn More

1-Methylindole-2-boronic acid, 95%, Thermo Scientific Chemicals

Catalog Number p-7035314
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
191162-40-0
C9H10BNO2
174.99
MFCD01114668
CBPBJUTWVXLSER-UHFFFAOYSA-N
1-methyl-1h-indol-2-yl boronic acid, n-methylindole-2-boronic acid, 2-borono-1-methyl-1h-indole, 1-methylindol-2-yl boronic acid, 1-methyl-1h-indole-2-boronic acid, 1-methyl-1h-indol-2-yl-2-boronic acid, boronic acid,b-1-methyl-1h-indol-2-yl, boronic acid, 1-methyl-1h-indol-2-yl, acmc-209eur, n-methyl-indoleboronic acid
22733820
CN1C(=CC2=CC=CC=C12)B(O)O
This item is not returnable. View return policy
191162-40-0
C9H10BNO2
174.99
MFCD01114668
CBPBJUTWVXLSER-UHFFFAOYSA-N
1-methyl-1h-indol-2-yl boronic acid, n-methylindole-2-boronic acid, 2-borono-1-methyl-1h-indole, 1-methylindol-2-yl boronic acid, 1-methyl-1h-indole-2-boronic acid, 1-methyl-1h-indol-2-yl-2-boronic acid, boronic acid,b-1-methyl-1h-indol-2-yl, boronic acid, 1-methyl-1h-indol-2-yl, acmc-209eur, n-methyl-indoleboronic acid
22733820
CN1C(=CC2=CC=CC=C12)B(O)O

Reactant for Suzuki-Miyaura coupling. It ia an active Pharmaceutical ingredient hence used as pharmaceutical intermediates. Also used as rawv material in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant for Suzuki-Miyaura coupling. It ia an active Pharmaceutical ingredient hence used as pharmaceutical intermediates. Also used as rawv material in organic synthesis.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at 4°C.
TRUSTED_SUSTAINABILITY
Melting Point 108°C to 112°C
Quantity 250 mg
IUPAC Name (1-methyl-1H-indol-2-yl)boronic acid
Formula Weight 174.99
Percent Purity 95%
Chemical Name or Material 1-Methylindole-2-boronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'