missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-imidazolemethanol, 98%, Thermo Scientific Chemicals

Catalog Number p-7055428
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
17334-08-6
C5H8N2O
112.132
MFCD00964673
CDQDMLWGTVLQEE-UHFFFAOYSA-N
1-methyl-1h-imidazol-2-yl methanol, 1-methylimidazol-2-yl methanol, 1h-imidazole-2-methanol, 1-methyl, 1-methyl-2-imidazolyl methanol, 2-hydroxymethyl-1-methylimidazole, imidazole-2-methanol, 1-methyl, 1-methyl-1h-imidazole-2-methanol, 1-methylimidazol-2-yl methan-1-ol, pubchem9047
573612
(1-methylimidazol-2-yl)methanol
CN1C=CN=C1CO
This item is not returnable. View return policy
17334-08-6
C5H8N2O
112.132
MFCD00964673
CDQDMLWGTVLQEE-UHFFFAOYSA-N
1-methyl-1h-imidazol-2-yl methanol, 1-methylimidazol-2-yl methanol, 1h-imidazole-2-methanol, 1-methyl, 1-methyl-2-imidazolyl methanol, 2-hydroxymethyl-1-methylimidazole, imidazole-2-methanol, 1-methyl, 1-methyl-1h-imidazole-2-methanol, 1-methylimidazol-2-yl methan-1-ol, pubchem9047
573612
(1-methylimidazol-2-yl)methanol
CN1C=CN=C1CO

1-Methyl-2-imidazolemethanol is used as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methyl-2-imidazolemethanol is used as intermediates.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acid, acid chlorides, halogens, halocarbons.
TRUSTED_SUSTAINABILITY
Melting Point 107°C to 111°C
Quantity 5 g
Beilstein 113492
Solubility Information Slightly soluble in water.
Formula Weight 112.13
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 1-Methyl-2-imidazolemethanol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'