missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-indole-5-carboxylic acid, 95%, Thermo Scientific™

Catalog Number p-3175480
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
186129-25-9
C10H9NO2
175.187
MFCD03839859
UHQAIJFIXCOBCN-UHFFFAOYSA-N
1-methyl-1h-indole-5-carboxylic acid, 1-methyl-5-indolecarboxylic acid, 1h-indole-5-carboxylic acid, 1-methyl, 1-methyl-1h-indole-5-carboxylicacid, 5-carboxy-1-methyl-1h-indole, 1h-indole-5-carboxylicacid,1-methyl, n-methylindol-5-carboxylic acid, 1-methyl-5-indolecarboxylicacid, methyl 1h-indole-5-carboxylic acid, 1-methyl-1h-indol-5-carboxylic acid
7015601
1-methylindole-5-carboxylic acid
CN1C=CC2=C1C=CC(=C2)C(=O)O
This item is not returnable. View return policy
186129-25-9
C10H9NO2
175.187
MFCD03839859
UHQAIJFIXCOBCN-UHFFFAOYSA-N
1-methyl-1h-indole-5-carboxylic acid, 1-methyl-5-indolecarboxylic acid, 1h-indole-5-carboxylic acid, 1-methyl, 1-methyl-1h-indole-5-carboxylicacid, 5-carboxy-1-methyl-1h-indole, 1h-indole-5-carboxylicacid,1-methyl, n-methylindol-5-carboxylic acid, 1-methyl-5-indolecarboxylicacid, methyl 1h-indole-5-carboxylic acid, 1-methyl-1h-indol-5-carboxylic acid
7015601
1-methylindole-5-carboxylic acid
CN1C=CC2=C1C=CC(=C2)C(=O)O
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 175.19
Percent Purity 95%
Chemical Name or Material 1-Methyl-1H-indole-5-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'