missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Indoleacetic acid, 95%

Brand:  Alfa Aesar™ H63935.06

Additional Details : CAS Number : 24297-59-4 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15430919

  • 11400.00 NOK / 5g


Estimated Shipment date
from Supplier 15-10-2019
Add to basket

Specifications

Chemical Identifiers

24297-59-4
175.187
WQJFIWXYPKYBTO-UHFFFAOYSA-N
90448
2-indol-1-ylacetic acid
C10H9NO2
MFCD00047262
1h-indole-1-acetic acid, 2-1h-indol-1-yl acetic acid, indol-1-yl-acetic acid, indole-1-acetic acid, 1h-indol-1-ylacetic acid, indol-1-ylacetic acid, indole-n-acetic acid, 1-indoleacetic acid, pubchem20663, 1-indolyl acetic acid
CHEBI:72814
C1=CC=C2C(=C1)C=CN2CC(=O)O

Specifications

5g
24297-59-4
MFCD00047262
1h-indole-1-acetic acid, 2-1h-indol-1-yl acetic acid, indol-1-yl-acetic acid, indole-1-acetic acid, 1h-indol-1-ylacetic acid, indol-1-ylacetic acid, indole-n-acetic acid, 1-indoleacetic acid, pubchem20663, 1-indolyl acetic acid
C1=CC=C2C(=C1)C=CN2CC(=O)O
175.187
CHEBI:72814
Solid
1-Indoleacetic acid
C10H9NO2
142924
WQJFIWXYPKYBTO-UHFFFAOYSA-N
2-indol-1-ylacetic acid
90448
175.18
95%
Videos
SDS Documents