missing translation for 'onlineSavingsMsg'
Learn More

1-Eicosanol, 98%, Thermo Scientific Chemicals

Catalog Number p-3757796
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
629-96-9
C20H42O
298.56
MFCD00002938
BTFJIXJJCSYFAL-UHFFFAOYSA-N
1-eicosanol, arachidyl alcohol, n-eicosanol, arachidic alcohol, arachic alcohol, eicosyl alcohol, eicosan-1-ol, eicosanol, 1-icosanol, n-1-eicosanol
12404
CHEBI:75627
icosan-1-ol
CCCCCCCCCCCCCCCCCCCCO
This item is not returnable. View return policy
629-96-9
C20H42O
298.56
MFCD00002938
BTFJIXJJCSYFAL-UHFFFAOYSA-N
1-eicosanol, arachidyl alcohol, n-eicosanol, arachidic alcohol, arachic alcohol, eicosyl alcohol, eicosan-1-ol, eicosanol, 1-icosanol, n-1-eicosanol
12404
CHEBI:75627
icosan-1-ol
CCCCCCCCCCCCCCCCCCCCO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 64°C to 66°C
Color White
Flash Point 194°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)19OH
Quantity 5 g
Beilstein 01, 431
Solubility Information Solubility in water: slightly soluble
Formula Weight 298.54
Percent Purity 98%
Physical Form Pellets, Powder or Chunks
Chemical Name or Material 1-Eicosanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'