missing translation for 'onlineSavingsMsg'
Learn More

1-Chloropentane, 98%, Thermo Scientific Chemicals

Catalog Number p-7027740
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
543-59-9
C5H11Cl
106.59
MFCD00001015
SQCZQTSHSZLZIQ-UHFFFAOYSA-N
pentyl chloride, pentane, 1-chloro, n-amyl chloride, n-pentyl chloride, amyl chloride, chloropentane, unii-0eg9msd3nk, 0eg9msd3nk, pentane, chloro, n-butylcarbonyl chloride
10977
1-chloropentane
CCCCCCl
This item is not returnable. View return policy
543-59-9
C5H11Cl
106.59
MFCD00001015
SQCZQTSHSZLZIQ-UHFFFAOYSA-N
pentyl chloride, pentane, 1-chloro, n-amyl chloride, n-pentyl chloride, amyl chloride, chloropentane, unii-0eg9msd3nk, 0eg9msd3nk, pentane, chloro, n-butylcarbonyl chloride
10977
1-chloropentane
CCCCCCl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -99°C
Density 0.881
Boiling Point 106°C to 108°C
Flash Point 3°C (37°F)
Odor Sweet
Linear Formula CH3(CH2)4Cl
Refractive Index 1.4125
Quantity 50 g
UN Number UN1107
Merck Index 14,606
Formula Weight 106.6
Percent Purity 98%
Chemical Name or Material 1-Chloropentane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'