missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-4-fluorobenzene, 98%, Thermo Scientific Chemicals

Catalog Number p-7021380
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
352-33-0
C6H4ClF
130.55
MFCD00000603
RJCGZNCCVKIBHO-UHFFFAOYSA-N
p-chlorofluorobenzene, p-fluorochlorobenzene, 4-chlorofluorobenzene, benzene, 1-chloro-4-fluoro, 1-fluoro-4-chlorobenzene, 1-chloro-4-fluoro-benzene, 4-fluorochlorobenzene, 1,4-fluorochlorobenzene, p-chloro-fluorobenzene, pubchem3420
9604
1-chloro-4-fluorobenzene
FC1=CC=C(Cl)C=C1
This item is not returnable. View return policy
352-33-0
C6H4ClF
130.55
MFCD00000603
RJCGZNCCVKIBHO-UHFFFAOYSA-N
p-chlorofluorobenzene, p-fluorochlorobenzene, 4-chlorofluorobenzene, benzene, 1-chloro-4-fluoro, 1-fluoro-4-chlorobenzene, 1-chloro-4-fluoro-benzene, 4-fluorochlorobenzene, 1,4-fluorochlorobenzene, p-chloro-fluorobenzene, pubchem3420
9604
1-chloro-4-fluorobenzene
FC1=CC=C(Cl)C=C1

1-Chloro-4-fluorobenzeneis used as a primary and secondary intermediates, organic synthesis. And it is also used to prepare (4'-fluoro-biphenyl-4-yl)-methyl ether by using reagents NaH, Ni (OAc)2, 2,2 '-bipyridyl, KI and solvents benzene, tetrahydrofuran at temperature of 63°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-4-fluorobenzeneis used as a primary and secondary intermediates, organic synthesis. And it is also used to prepare (4′-fluoro-biphenyl-4-yl)-methyl ether by using reagents NaH, Ni (OAc)2, 2,2 ′-bipyridyl, KI and solvents benzene, tetrahydrofuran at temperature of 63°C.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Melting Point -22°C
Density 1.228
Boiling Point 129°C to 130°C
Flash Point 29°C (84°F)
Refractive Index 1.495
Quantity 25 g
UN Number UN1993
Beilstein 1904542
Solubility Information Difficult to mix.
Formula Weight 130.55
Percent Purity 98%
Chemical Name or Material 1-Chloro-4-fluorobenzene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'