missing translation for 'onlineSavingsMsg'
Learn More

1-Bromononane, 99%, Thermo Scientific Chemicals

Catalog Number p-7080854
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
693-58-3
C9H19Br
207.155
MFCD00000278
AYMUQTNXKPEMLM-UHFFFAOYSA-N
n-nonyl bromide, nonyl bromide, nonane, 1-bromo, 1-nonyl bromide, 1-n-nonyl bromide, n-nonyl-1-bromide, bromononane, nonylbromide, 1-bromo-n-nonane, labotest-bb ltbb001163
12742
1-bromononane
CCCCCCCCCBr
This item is not returnable. View return policy
693-58-3
C9H19Br
207.155
MFCD00000278
AYMUQTNXKPEMLM-UHFFFAOYSA-N
n-nonyl bromide, nonyl bromide, nonane, 1-bromo, 1-nonyl bromide, 1-n-nonyl bromide, n-nonyl-1-bromide, bromononane, nonylbromide, 1-bromo-n-nonane, labotest-bb ltbb001163
12742
1-bromononane
CCCCCCCCCBr

1-Bromononane is used as organic intermediates. It is mainly used in pharmaceutical synthesis and industrial dye intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromononane is used as organic intermediates. It is mainly used in pharmaceutical synthesis and industrial dye intermediates.

Solubility
Insoluble in water. Soluble in ethyl ether and benzene.

Notes
Stable under normal conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 1.086
Boiling Point 200°C to 201°C
Flash Point 90°C (194°F)
Refractive Index 1.455
Quantity 25 g
Beilstein 1733632
Solubility Information Insoluble in water. Soluble in ethyl ether and benzene.
Formula Weight 207.16
Percent Purity 99%
Chemical Name or Material 1-Bromononane
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 1
Hazard Statement
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'