missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-4-cyclohexylbenzene, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
1-Bromo-4-cyclohexylbenzene is used as a organic light-emitting diode and as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Bromo-4-cyclohexylbenzene is used as a organic light-emitting diode and as a pharmaceutical intermediate.
Solubility
Not miscible or difficult to mix with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
1-Bromo-4-cyclohexylbenzene is used as a organic light-emitting diode and as a pharmaceutical intermediate.
Solubility
Not miscible or difficult to mix with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Density | 1.287 |
Boiling Point | 90°C to 94°C (0.01 mmHg) |
Refractive Index | 1.5595 |
Quantity | 5 g |
Beilstein | 2209869 |
Solubility Information | Not miscible or difficult to mix with water. |
Formula Weight | 239.16 |
Percent Purity | 98% |
Chemical Name or Material | 1-Bromo-4-cyclohexylbenzene |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'