missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Boc-indoline-5-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
1-Boc-indoline-5-boronic acid pinacol ester is used as disinfector, tanning agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Boc-indoline-5-boronic acid pinacol ester is used as disinfector, tanning agent.
Solubility
Soluble in dichloromethane, ethyl acetate and methanol.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
1-Boc-indoline-5-boronic acid pinacol ester is used as disinfector, tanning agent.
Solubility
Soluble in dichloromethane, ethyl acetate and methanol.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Quantity | 250 mg |
Solubility Information | Soluble in dichloromethane,ethyl acetate and methanol. |
Formula Weight | 345.25 |
Percent Purity | 97% |
Chemical Name or Material | 1-Boc-indoline-5-boronic acid pinacol ester |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'