missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Boc-6-bromoindole, 97%

Brand:  Alfa Aesar™ H63538.06

Additional Details : CAS Number : 147621-26-9 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15430279

  • 6362.73 NOK / 5g


Estimated Shipment
09-09-2020
Add to basket

Specifications

Chemical Identifiers

147621-26-9
296.164
OUOWFTDOMYJYKB-UHFFFAOYSA-N
11449321
CC(C)(C)OC(=O)N1C=CC2=C1C=C(C=C2)Br
C13H14BrNO2
MFCD09954826
tert-butyl 6-bromo-1h-indole-1-carboxylate, 1-boc-6-bromoindole, acmc-209cyx, 1-tert-butoxycarbonyl-6-bromoindole, tert-butyl6-bromo-1h-indole-1-carboxylate, n-tert-butoxycarbonyl-6-bromoindole, 1,1-dimethylethyl 6-bromo-1h-indole-1-carboxylate, 6-bromo-1h-indole-1-carboxylic acid tert-butyl ester, 6-bromo-indole-1-carboxylic acid tert-butyl ester, 6-bromo-1h-indole-1-carboxylic acid 1,1-dimethylethyl ester
tert-butyl 6-bromoindole-1-carboxylate

Specifications

1-Boc-6-bromoindole
147621-26-9
MFCD09954826
tert-butyl 6-bromo-1h-indole-1-carboxylate, 1-boc-6-bromoindole, acmc-209cyx, 1-tert-butoxycarbonyl-6-bromoindole, tert-butyl6-bromo-1h-indole-1-carboxylate, n-tert-butoxycarbonyl-6-bromoindole, 1,1-dimethylethyl 6-bromo-1h-indole-1-carboxylate, 6-bromo-1h-indole-1-carboxylic acid tert-butyl ester, 6-bromo-indole-1-carboxylic acid tert-butyl ester, 6-bromo-1h-indole-1-carboxylic acid 1,1-dimethylethyl ester
CC(C)(C)OC(=O)N1C=CC2=C1C=C(C=C2)Br
296.164
296.16
97%
5g
C13H14BrNO2
9050272
OUOWFTDOMYJYKB-UHFFFAOYSA-N
tert-butyl 6-bromoindole-1-carboxylate
11449321
Solid
Videos
SDS Documents