missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Boc-5-nitroindole, 97%

Brand:  Alfa Aesar™ H63394.06

Additional Details : CAS Number : 166104-19-4 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15430069

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Specifications

Chemical Identifiers

166104-19-4
262.265
GSHXPDVMVVLYLH-UHFFFAOYSA-N
11680504
CC(C)(C)OC(=O)N1C=CC2=C1C=CC(=C2)[N+](=O)[O-]
C13H14N2O4
MFCD09953094
tert-butyl 5-nitro-1h-indole-1-carboxylate, 1-boc-5-nitro-1h-indole, pubchem16753, tert-butyl 5-nitro-indole-1-carboxylate, tert-butyl-5-nitro-1h-indole-1-carboxylate, 5-nitro-1-indolecarboxylic acid tert-butyl ester, 5-nitro-indole-1-carboxylic acid tert-butyl ester, 5-nitro-1h-indole-1-carboxylic acid tert-butyl ester, 1h-indole-1-carboxylicacid,5-nitro-,1,1-dimethylethyl ester
tert-butyl 5-nitroindole-1-carboxylate

Specifications

1-Boc-5-nitroindole
166104-19-4
MFCD09953094
tert-butyl 5-nitro-1h-indole-1-carboxylate, 1-boc-5-nitro-1h-indole, pubchem16753, tert-butyl 5-nitro-indole-1-carboxylate, tert-butyl-5-nitro-1h-indole-1-carboxylate, 5-nitro-1-indolecarboxylic acid tert-butyl ester, 5-nitro-indole-1-carboxylic acid tert-butyl ester, 5-nitro-1h-indole-1-carboxylic acid tert-butyl ester, 1h-indole-1-carboxylicacid,5-nitro-,1,1-dimethylethyl ester
CC(C)(C)OC(=O)N1C=CC2=C1C=CC(=C2)[N+](=O)[O-]
262.265
262.26
97%
5g
C13H14N2O4
10344204
GSHXPDVMVVLYLH-UHFFFAOYSA-N
tert-butyl 5-nitroindole-1-carboxylate
11680504
Solid
Videos
SDS Documents