missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Boc-3-hydroxypiperidine, 97%, Thermo Scientific Chemicals
380.41 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
Description
It is a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a pharmaceutical intermediate.
Notes
Store at room temperature. Keep away from strong oxidizing agents. Ensure proper ventilation.
It is a pharmaceutical intermediate.
Notes
Store at room temperature. Keep away from strong oxidizing agents. Ensure proper ventilation.
Specifications
Melting Point | 66°C to 70°C |
Quantity | 1 g |
Beilstein | 4180859 |
Formula Weight | 201.27 |
Percent Purity | 97% |
Chemical Name or Material | 1-Boc-3-hydroxypiperidine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'