missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+/-)-1-Boc-3-bromopyrrolidine, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
(±)-1-Boc-3-bromopyrrolidine is used in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(±)-1-Boc-3-bromopyrrolidine is used in organic synthesis.
Solubility
Slightly soluble in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
(±)-1-Boc-3-bromopyrrolidine is used in organic synthesis.
Solubility
Slightly soluble in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications
Melting Point | 45°C to 53°C |
Quantity | 250 mg |
UN Number | UN3077 |
Synonym | n-boc-3-bromopyrrolidine, 1-boc-3-bromopyrrolidine, 3-bromo-pyrrolidine-1-carboxylic acid tert-butyl ester, +/--1-boc-3-bromopyrrolidine, acmc-209zg9, ^+-1-boc-3-bromopyrrolidine, rs 1-tertbutoxycarbonyl-3-bromopyrrolidine, rs 1-tert-butoxycarbonyl-3-bromopyrrolidine, 2-methyl-2-propanyl 3-bromo-1-pyrrolidinecarboxylate |
Solubility Information | Slightly soluble in water. |
Formula Weight | 250.14 |
Percent Purity | 95% |
Chemical Name or Material | (+/-)-1-Boc-3-bromopyrrolidine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'