missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 1-Boc-3,3-difluoropyrrolidine, 98%
Click to view available options
Quantity:
250 mg
1 g
Specifications
Refractive Index | 1.42 |
Quantity | 250 mg |
Formula Weight | 207.22 |
Percent Purity | 98% |
Chemical Name or Material | 1-Boc-3,3-difluoropyrrolidine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'