missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Benzenesulfonyl-5-cyanoindole, 97%

Brand:  Alfa Aesar™ H63313.MD

Additional Details : CAS Number : 144340-21-6 Weight : 0.00025kg Transport : UN number : 3439 Chem class : 6.1 Pack group : III

Quantity 250mg

 View more versions of this product

Product Code. 15429948

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Specifications

Chemical Identifiers

144340-21-6
282.317
JAQJCWQKPPYZCW-UHFFFAOYSA-N
15289975
C1=CC=C(C=C1)S(=O)(=O)N2C=CC3=C2C=CC(=C3)C#N
C15H10N2O2S
MFCD09965529
1-benzenesulfonyl-1h-indole-5-carbonitrile, 1-benzenesulfonyl indole-5-carbonitrile, 1-phenylsulfonyl-1h-indole-5-carbonitrile
1-(benzenesulfonyl)indole-5-carbonitrile

Specifications

1-Benzenesulfonyl-5-cyanoindole
250mg
C15H10N2O2S
UN3439
1-benzenesulfonyl-1h-indole-5-carbonitrile, 1-benzenesulfonyl indole-5-carbonitrile, 1-phenylsulfonyl-1h-indole-5-carbonitrile
C1=CC=C(C=C1)S(=O)(=O)N2C=CC3=C2C=CC(=C3)C#N
282.317
282.32
97%
White
144340-21-6
MFCD09965529
7644292
JAQJCWQKPPYZCW-UHFFFAOYSA-N
1-(benzenesulfonyl)indole-5-carbonitrile
15289975
Solid
Videos
SDS Documents