missing translation for 'onlineSavingsMsg'
Learn More

1-Amino-4-oxocyclohexanecarboxylic acid ethylene ketal, 98%, Thermo Scientific Chemicals

Catalog Number p-7047813
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
54621-18-0
C9H15NO4
201.222
MFCD03790906
HIIFALCQUKTUHB-UHFFFAOYSA-N
8-amino-1,4-dioxaspiro 4.5 decane-8-carboxylic acid, 1-amino-4-oxocyclohexanecarboxylic acid ethylene ketal, 1-amino-4-oxocyclohexanecarboxylicacidethyleneketal, 1-amino-4,4-ethylenedioxycyclohexane carboxylic acid, 4-amino-4-carboxycyclohexanone ethylene ketal, 1,4-dioxaspiro 4.5 decane-8-carboxylicacid, 8-amino, 4-amino-4-2-hydroxy-1,3-dioxolan-2-yl cyclohexanone, 8-amino-1,4-dioxa-spiro 4.5 decane-8-carboxylic acid, amino-4-ethylene-ketal cyclohexylcarboxylic acid, 1-amino-4-oxocyclohexanecarboxylic acid ethylene k
7015505
8-amino-1,4-dioxaspiro[4.5]decane-8-carboxylic acid
C1CC2(CCC1(C(=O)O)N)OCCO2
This item is not returnable. View return policy
54621-18-0
C9H15NO4
201.222
MFCD03790906
HIIFALCQUKTUHB-UHFFFAOYSA-N
8-amino-1,4-dioxaspiro 4.5 decane-8-carboxylic acid, 1-amino-4-oxocyclohexanecarboxylic acid ethylene ketal, 1-amino-4-oxocyclohexanecarboxylicacidethyleneketal, 1-amino-4,4-ethylenedioxycyclohexane carboxylic acid, 4-amino-4-carboxycyclohexanone ethylene ketal, 1,4-dioxaspiro 4.5 decane-8-carboxylicacid, 8-amino, 4-amino-4-2-hydroxy-1,3-dioxolan-2-yl cyclohexanone, 8-amino-1,4-dioxa-spiro 4.5 decane-8-carboxylic acid, amino-4-ethylene-ketal cyclohexylcarboxylic acid, 1-amino-4-oxocyclohexanecarboxylic acid ethylene k
7015505
8-amino-1,4-dioxaspiro[4.5]decane-8-carboxylic acid
C1CC2(CCC1(C(=O)O)N)OCCO2

1-Amino-4-oxocyclohexanecarboxylic acid ethylene ketal is used in modification of polymer properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Amino-4-oxocyclohexanecarboxylic acid ethylene ketal is used in modification of polymer properties.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 301°C to 304°C
Quantity 250 mg
Beilstein 1373325
Solubility Information Insoluble in water.
Formula Weight 201.22
Percent Purity 98%
Chemical Name or Material 1-Amino-4-oxocyclohexanecarboxylic acid ethylene ketal

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'