Learn More
1-Allylimidazole, 97%, Thermo Scientific Chemicals
Catalog Number p-51811
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
31410-01-2
C6H8N2
108.14
MFCD00037894
XLXCHZCQTCBUOX-UHFFFAOYSA-N
1-allylimidazole, 1-allyl-1h-imidazole, 1-prop-2-en-1-yl-1h-imidazole, 1h-imidazole, 1-2-propenyl, 1-prop-2-en-1-yl imidazole, imidazole, 1-allyl, n-allylimidazole, 1-allyl imidazole, acmc-1cr2q, 1-allyl-1h-imidazole #
35794
1-prop-2-enylimidazole
C=CCN1C=CN=C1
31410-01-2
C6H8N2
108.14
MFCD00037894
XLXCHZCQTCBUOX-UHFFFAOYSA-N
1-allylimidazole, 1-allyl-1h-imidazole, 1-prop-2-en-1-yl-1h-imidazole, 1h-imidazole, 1-2-propenyl, 1-prop-2-en-1-yl imidazole, imidazole, 1-allyl, n-allylimidazole, 1-allyl imidazole, acmc-1cr2q, 1-allyl-1h-imidazole #
35794
1-prop-2-enylimidazole
C=CCN1C=CN=C1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Yellow to Orange
Density 1.0000g/mL
Boiling Point 212.0°C to 213.0°C
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5030 to 1.5050
Quantity 25 g
Specific Gravity 1
Formula Weight 108.14
Percent Purity 97%
Physical Form Viscous Liquid
Chemical Name or Material 1-Allylimidazole, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Allylimidazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.