missing translation for 'onlineSavingsMsg'
Learn More

1-Adamantaneacetic acid, 98+%, Thermo Scientific Chemicals

Catalog Number p-7045800
283.21 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
4942-47-6
C12H17O2
193.27
MFCD00074728
AOTQGWFNFTVXNQ-UHFFFAOYSA-M
1-adamantaneacetic acid, 2-adamantan-1-yl acetic acid, 1-adamantylacetic acid, 1-adamantane acetic acid, adamantan-1-ylacetic acid, 2-1-adamantyl acetic acid, tricyclo 3.3.1.13,7 dec-1-ylacetic acid, adamantan-1-yl-acetic acid, 1-carboxymethyl adamantane
123221
2-(1-adamantyl)acetic acid
[O-]C(=O)CC12CC3CC(CC(C3)C1)C2
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
4942-47-6
C12H17O2
193.27
MFCD00074728
AOTQGWFNFTVXNQ-UHFFFAOYSA-M
1-adamantaneacetic acid, 2-adamantan-1-yl acetic acid, 1-adamantylacetic acid, 1-adamantane acetic acid, adamantan-1-ylacetic acid, 2-1-adamantyl acetic acid, tricyclo 3.3.1.13,7 dec-1-ylacetic acid, adamantan-1-yl-acetic acid, 1-carboxymethyl adamantane
123221
2-(1-adamantyl)acetic acid
[O-]C(=O)CC12CC3CC(CC(C3)C1)C2

1-Adamantaneacetic acid is used as an acylating agent in determining pharmacological characteristics of ten new analogues of bradykinin (Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg) that were modified in the N-terminal part of the molecule.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Adamantaneacetic acid is used as an acylating agent in determining pharmacological characteristics of ten new analogues of bradykinin (Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg) that were modified in the N-terminal part of the molecule.

Solubility
Solubility in Methanol - almost transparency

Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place
TRUSTED_SUSTAINABILITY
Melting Point 136°C to 138°C
Quantity 1 g
Beilstein 641412
Formula Weight 194.28
Percent Purity ≥98%
Chemical Name or Material 1-Adamantaneacetic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'