Learn More
1-Acetylpiperazine, 98%
Description
1-Acetylpiperazine may be used in the synthesis of series of 7-alkoxyl substituted indolizinoquinoline-5,12-dione derivatives and 2-substituted-N-(naphth-1-ylmethyl)-pyrimidin-4-amines and 2-substituted-N-benzhydrylpyrimidin-4-amines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Acetylpiperazine may be used in the synthesis of series of 7-alkoxyl substituted indolizinoquinoline-5,12-dione derivatives and 2-substituted-N-(naphth-1-ylmethyl)-pyrimidin-4-amines and 2-substituted-N-benzhydrylpyrimidin-4-amines.
Solubility
Soluble in water 210 g/L (20°C).
Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, moisture.
Specifications
Specifications
| Melting Point | 30°C to 34°C |
| Boiling Point | 127°C to 130°C (8 mmHg) |
| Flash Point | >110°C |
| Quantity | 10 g |
| Beilstein | 112220 |
| Sensitivity | Hygroscopic |
| Solubility Information | Soluble aq. soln. 210 g/L (20°C). |
| Formula Weight | 128.18 |
| Percent Purity | 99% |
| Chemical Name or Material | 1-Acetylpiperazine |
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.