missing translation for 'onlineSavingsMsg'
Learn More

1-Acetyl-4-(4-hydroxyphenyl)piperazine, 98%, Thermo Scientific™

Catalog Number p-7026209
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
67914-60-7
C12H16N2O2
220.27
MFCD00044905
AGVNLFCRZULMKK-UHFFFAOYSA-N
1-acetyl-4-4-hydroxyphenyl piperazine, 4-1-acetylpiperazin-4-yl phenol, 1-4-4-hydroxyphenyl piperazin-1-yl ethanone, n-acetyl-4-4-hydroxyphenyl piperazine, 4-4-acetyl-1-piperazinyl phenol, piperazine, 1-acetyl-4-4-hydroxyphenyl, 4-4-acetylpiperazin-4-yl phenol, 1-4-4-hydroxyphenyl piperazin-1-yl ethan-1-one, 1-acetyl-4-4-hydroxylphenyl piperazine
712441
1-[4-(4-hydroxyphenyl)piperazin-1-yl]ethanone
CC(=O)N1CCN(CC1)C1=CC=C(O)C=C1
This item is not returnable. View return policy
67914-60-7
C12H16N2O2
220.27
MFCD00044905
AGVNLFCRZULMKK-UHFFFAOYSA-N
1-acetyl-4-4-hydroxyphenyl piperazine, 4-1-acetylpiperazin-4-yl phenol, 1-4-4-hydroxyphenyl piperazin-1-yl ethanone, n-acetyl-4-4-hydroxyphenyl piperazine, 4-4-acetyl-1-piperazinyl phenol, piperazine, 1-acetyl-4-4-hydroxyphenyl, 4-4-acetylpiperazin-4-yl phenol, 1-4-4-hydroxyphenyl piperazin-1-yl ethan-1-one, 1-acetyl-4-4-hydroxylphenyl piperazine
712441
1-[4-(4-hydroxyphenyl)piperazin-1-yl]ethanone
CC(=O)N1CCN(CC1)C1=CC=C(O)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 180°C to 185°C
Quantity 25 g
Beilstein 795777
Formula Weight 220.27
Percent Purity 98%
Chemical Name or Material 1-Acetyl-4-(4-hydroxyphenyl)piperazine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'