missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,5-Dihydroxynaphthalene, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1000 g
Description
1,5-Dihydroxynaphthalene is an intermediate of synthetic mordant azo dyes. It is an intermediate used in organic synthesis, pharmaceuticals, dyestuff fields and photograph industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,5-Dihydroxynaphthalene is an intermediate of synthetic mordant azo dyes. It is an intermediate used in organic synthesis, pharmaceuticals, dyestuff fields and photograph industry.
Solubility
Soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
1,5-Dihydroxynaphthalene is an intermediate of synthetic mordant azo dyes. It is an intermediate used in organic synthesis, pharmaceuticals, dyestuff fields and photograph industry.
Solubility
Soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Melting Point | ∽260°C (decomposition) |
Flash Point | 252°C (486°F) |
Quantity | 250 g |
Beilstein | 2044951 |
Solubility Information | Soluble in water. |
Formula Weight | 160.17 |
Percent Purity | 98% |
Chemical Name or Material | 1,5-Dihydroxynaphthalene |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'