Learn More
Thermo Scientific™ 1.5/2mL Microtube Adapter
Accommodate 1.5/2mL microtubes in the Thermo Scientific™ MicroClick 18 x 5 rotor.
Brand: Thermo Scientific™ 75005756
Additional Details : Weight : 0.20000kg
Description
1.5/2mL adapters for use with the Thermo Scientific MicroClick 18 x 5 rotor.
Specifications
Microtube Adapter | |
18 x 1.5/2 mL | |
11 mm | |
1.5/2mL Microtube Adapter |
Microtube | |
45 mm | |
MicroClick 18 x 5 rotor | |
1.5/2 mL |