Learn More
1,4-Diiodobenzene, 98%, Thermo Scientific Chemicals
Use promo code "21615" to get your promotional price.
Description
1,4-Diiodobenzene is used in Suzuki reaction. It is used as a precursor in the preparation of martinellic acid and 1,4-bis(p-R-phenylethynyl)benzenes. Further, it is employed in the preparation of 1,4-diiodo-2,5-didodecylbenzene , which is a starting reagent for the synthesis of oligo(1,4-phenylene ethynylene). It is also used in human and animal nutrition products including antiseptics and disinfectants. In addition to this, it used in polarizing films for liquid crystal display chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,4-Diiodobenzene is used in Suzuki reaction. It is used as a precursor in the preparation of martinellic acid and 1,4-bis(p-R-phenylethynyl)benzenes. Further, it is employed in the preparation of 1,4-diiodo-2,5-didodecylbenzene , which is a starting reagent for the synthesis of oligo(1,4-phenylene ethynylene). It is also used in human and animal nutrition products including antiseptics and disinfectants. In addition to this, it used in polarizing films for liquid crystal display chemicals.
Solubility
Soluble in ether and ethanol. Insoluble in water.
Notes
Light sensitive. Incompatible with oxidizing agents.
Specifications
Melting Point | 128°C to 132°C |
Boiling Point | 285°C |
Quantity | 25 g |
Beilstein | 1904546 |
Sensitivity | Light sensitive |
Solubility Information | Soluble in ether and ethanol. Insoluble in water. |
Formula Weight | 329.91 |
Percent Purity | 98% |
Chemical Name or Material | 1,4-Diiodobenzene |
RUO – Research Use Only