missing translation for 'onlineSavingsMsg'
Learn More

1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(2,4-difluorophenyl)urea, 97%, Alfa Aesar™

Brand:  Alfa Aesar H65383.MD

Additional Details : Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15429606

  • 890.00 NOK / 250mg


Estimated Shipment date
from Supplier 21-11-2019
Add to basket

Specifications

Chemical Identifiers

793677-93-7
350.67
CUXRGCFDGKWHAO-UHFFFAOYSA-N
2554140
C14H8ClF5N2O
MFCD06363378
1-4-chloro-3-trifluoromethyl phenyl-3-2,4-difluorophenyl urea

Specifications

250mg
793677-93-7
MFCD06363378
CUXRGCFDGKWHAO-UHFFFAOYSA-N
2554140
Solid
1-[4-Chloro-3-(trifluoromethyl)phenyl]-3-(2,4-difluorophenyl)urea
C14H8ClF5N2O
1-4-chloro-3-trifluoromethyl phenyl-3-2,4-difluorophenyl urea
350.67
350.67
97%
Videos
SDS Documents