missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Chloro-2-nitrophenyl)homopiperazine, 96%

Brand:  Alfa Aesar™ H63766.03

Additional Details : CAS Number : 443923-21-5 Weight : 0.00100kg Transport : UN number : 3259 Chem class : 8 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15430629

  • 1911.50 NOK / 1g


Estimated Shipment
05-08-2020
Add to basket

Specifications

Chemical Identifiers

443923-21-5
1-(4-Chloro-2-nitrophenyl)-1,4-diazepane

Specifications

1-(4-Chloro-2-nitrophenyl)homopiperazine
443923-21-5
1-(4-Chloro-2-nitrophenyl)-1,4-diazepane
Solid
1g
UN3259
255.7
96%
Videos
SDS Documents