Learn More
1-(4-Chloro-2-nitrophenyl)homopiperazine, 96%, Thermo Scientific™
Catalog Number 15450629
Click to view available options
Quantity:
250 mg
1 g
5 g
443923-21-5
1-(4-Chloro-2-nitrophenyl)-1,4-diazepane
443923-21-5
1-(4-Chloro-2-nitrophenyl)-1,4-diazepane
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN3259
Formula Weight 255.7
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 1-(4-Chloro-2-nitrophenyl)homopiperazine