missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Bromophenyl)octane, 97%, Thermo Scientific™

Catalog Number p-2694312
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
51554-93-9
C14H21Br
269.226
MFCD00038348
OOZQSVXPBCINJF-UHFFFAOYSA-N
1-4-bromophenyl octane, 1-bromo-4-n-octylbenzene, p-octylbromobenzene, p-bromophenyloctane, 4-n-octylbromobenzene, 1-p-bromophenyl octane, 4-bromooctylbenzene, 4-oct-1-yl bromobenzene, benzene, 1-bromo-4-octyl, 4-bromo-1-octylbenzene
142854
1-bromo-4-octylbenzene
CCCCCCCCC1=CC=C(C=C1)Br
This item is not returnable. View return policy
51554-93-9
C14H21Br
269.226
MFCD00038348
OOZQSVXPBCINJF-UHFFFAOYSA-N
1-4-bromophenyl octane, 1-bromo-4-n-octylbenzene, p-octylbromobenzene, p-bromophenyloctane, 4-n-octylbromobenzene, 1-p-bromophenyl octane, 4-bromooctylbenzene, 4-oct-1-yl bromobenzene, benzene, 1-bromo-4-octyl, 4-bromo-1-octylbenzene
142854
1-bromo-4-octylbenzene
CCCCCCCCC1=CC=C(C=C1)Br
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula BrC6H4(CH2)7CH3
Quantity 1 g
Formula Weight 269.22
Percent Purity 97%
Chemical Name or Material 1-(4-Bromophenyl)octane
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'