Learn More
(+/-)-1-(4-Bromophenyl)ethylamine, 96%, Thermo Scientific Chemicals
Catalog Number p-7045749
Click to view available options
Quantity:
1 g
5 g
24358-62-1
C8H10BrN
200.079
MFCD00025548
SOZMSEPDYJGBEK-UHFFFAOYSA-N
1-4-bromophenyl ethylamine, 1-4-bromophenyl ethanamine, 4-bromo-alpha-methylbenzylamine, 4-bromo-alpha-phenethylamine, 1-4-bromophenyl ethan-1-amine, 1-4-bromophenyl ethyl amine, --1-4-bromophenyl ethylamine, +-p-bromo-.alpha.-phenethylamine, 1-amino-1-4-bromophenyl ethane, +/--1-4-bromophenyl ethylamine
91175
1-(4-bromophenyl)ethanamine
CC(C1=CC=C(C=C1)Br)N
24358-62-1
C8H10BrN
200.079
MFCD00025548
SOZMSEPDYJGBEK-UHFFFAOYSA-N
1-4-bromophenyl ethylamine, 1-4-bromophenyl ethanamine, 4-bromo-alpha-methylbenzylamine, 4-bromo-alpha-phenethylamine, 1-4-bromophenyl ethan-1-amine, 1-4-bromophenyl ethyl amine, --1-4-bromophenyl ethylamine, +-p-bromo-.alpha.-phenethylamine, 1-amino-1-4-bromophenyl ethane, +/--1-4-bromophenyl ethylamine
91175
1-(4-bromophenyl)ethanamine
CC(C1=CC=C(C=C1)Br)N
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Density 1.397
Boiling Point 140°C to 145°C (30 mmHg)
Flash Point >110°C (230°F)
Odor Amine-like
Refractive Index 1.567
Quantity 1 g
UN Number UN2735
Beilstein 2412318
Sensitivity Air Sensitive
Formula Weight 200.08
Percent Purity 96%
Chemical Name or Material (+/-)-1-(4-Bromophenyl)ethylamine