Learn More
1,4-Benzodioxan-5-carboxylic acid, 98%, Thermo Scientific™
Catalog Number p-3770942
Click to view available options
Quantity:
1 g
5 g
4442-53-9
C9H8O4
180.16
MFCD00239415
VCLSWKVAHAJSFL-UHFFFAOYSA-N
1,4-benzodioxan-5-carboxylic acid, 2,3-dihydrobenzo b 1,4 dioxine-5-carboxylic acid, 2,3-dihydro-1,4-benzodioxin-5-carboxylic acid, 1,4-benzodioxane-5-carboxylic acid, 2,3-dihydro-benzo 1,4 dioxine-5-carboxylic acid, 2,3-ethylenedioxybenzoic acid, 2,3-dihydro-1,4-benzodioxane-5-carboxylic acid, 2,3-dihydrobenzo 1,4 dioxine-5-carboxylic acid, 1,4-benzodioxin-5-carboxylic acid, 2,3-dihydro, 2h,3h-benzo e 1,4-dioxin-5-carboxylic acid
78184
2,3-dihydro-1,4-benzodioxine-5-carboxylic acid
C1COC2=C(C=CC=C2O1)C(=O)O
4442-53-9
C9H8O4
180.16
MFCD00239415
VCLSWKVAHAJSFL-UHFFFAOYSA-N
1,4-benzodioxan-5-carboxylic acid, 2,3-dihydrobenzo b 1,4 dioxine-5-carboxylic acid, 2,3-dihydro-1,4-benzodioxin-5-carboxylic acid, 1,4-benzodioxane-5-carboxylic acid, 2,3-dihydro-benzo 1,4 dioxine-5-carboxylic acid, 2,3-ethylenedioxybenzoic acid, 2,3-dihydro-1,4-benzodioxane-5-carboxylic acid, 2,3-dihydrobenzo 1,4 dioxine-5-carboxylic acid, 1,4-benzodioxin-5-carboxylic acid, 2,3-dihydro, 2h,3h-benzo e 1,4-dioxin-5-carboxylic acid
78184
2,3-dihydro-1,4-benzodioxine-5-carboxylic acid
C1COC2=C(C=CC=C2O1)C(=O)O
Melting Point 90°C to 94°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 180.16
Percent Purity 98%
Chemical Name or Material 1, 4-Benzodioxan-5-carboxylic acid
compliance-icons
Product Identifier
  • 1,4-Benzodioxan-5-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.