Learn More
1,4-Benzodioxan-5-amine, 95%, Thermo Scientific Chemicals
Catalog Number p-4228354
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
16081-45-1
C8H9NO2
151.17
MFCD03695459
DMLRSJNZORFCBD-UHFFFAOYSA-N
5-amino-1,4-benzodioxane, 2,3-dihydrobenzo b 1,4 dioxin-5-amine, 2,3-dihydro-benzo 1,4 dioxin-5-ylamine, 2,3-ethylenedioxyaniline, 1,4-benzodioxin-5-amine,2,3-dihydro, 1,4-benzodioxan-5-amine, 5-amino-1,4-benzodioxan, 5-amino-1,4-dibenzodioxane, 1,4-benzodioxin-5-amine, 2,3-dihydro, 5-amino-2,3-dihydro-1,4-benzodioxin
11788387
2,3-dihydro-1,4-benzodioxin-5-amine
C1COC2=C(C=CC=C2O1)N
16081-45-1
C8H9NO2
151.17
MFCD03695459
DMLRSJNZORFCBD-UHFFFAOYSA-N
5-amino-1,4-benzodioxane, 2,3-dihydrobenzo b 1,4 dioxin-5-amine, 2,3-dihydro-benzo 1,4 dioxin-5-ylamine, 2,3-ethylenedioxyaniline, 1,4-benzodioxin-5-amine,2,3-dihydro, 1,4-benzodioxan-5-amine, 5-amino-1,4-benzodioxan, 5-amino-1,4-dibenzodioxane, 1,4-benzodioxin-5-amine, 2,3-dihydro, 5-amino-2,3-dihydro-1,4-benzodioxin
11788387
2,3-dihydro-1,4-benzodioxin-5-amine
C1COC2=C(C=CC=C2O1)N
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Color Yellow to Amber
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 151.17
Percent Purity 95%
Physical Form Beads
Chemical Name or Material 1, 4-Benzodioxan-5-amine
Show More Show Less
compliance-icons
Product Identifier
  • 1,4-Benzodioxan-5-amine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .