missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzenediboronic acid bis(neopentyl glycol) ester, 99%, Thermo Scientific™

Catalog Number p-7046748
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
5565-36-6
C16H24B2O4
301.984
MFCD01075726
XDODHEZHZCJSML-UHFFFAOYSA-N
1,4-benzenediboronic acid bis neopentyl glycol ester, 1,4-bis 5,5-dimethyl-1,3,2-dioxaborinan-2-yl benzene, 2-4-5,5-dimethyl-1,3,2-dioxaborinan-2-yl phenyl-5,5-dimethyl-1,3,2-dioxaborinane, 1,4-benzenediboronic acid bis npg cyclic ester, 1,4-benzenediboronic acid bisneopentylglycol cyclic ester, 1,4-bis 5,5-dimethyl-1,3,2-dioxaborinane-2-yl benzene, 1,4-benzenediboronic acid bis neopentyl glycol cyclic ester, 2,2'-1,4-phenylene bis 5,5-dimethyl-1,3,2-dioxaborinane
23005407
2-[4-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)phenyl]-5,5-dimethyl-1,3,2-dioxaborinane
B1(OCC(CO1)(C)C)C2=CC=C(C=C2)B3OCC(CO3)(C)C
This item is not returnable. View return policy
5565-36-6
C16H24B2O4
301.984
MFCD01075726
XDODHEZHZCJSML-UHFFFAOYSA-N
1,4-benzenediboronic acid bis neopentyl glycol ester, 1,4-bis 5,5-dimethyl-1,3,2-dioxaborinan-2-yl benzene, 2-4-5,5-dimethyl-1,3,2-dioxaborinan-2-yl phenyl-5,5-dimethyl-1,3,2-dioxaborinane, 1,4-benzenediboronic acid bis npg cyclic ester, 1,4-benzenediboronic acid bisneopentylglycol cyclic ester, 1,4-bis 5,5-dimethyl-1,3,2-dioxaborinane-2-yl benzene, 1,4-benzenediboronic acid bis neopentyl glycol cyclic ester, 2,2'-1,4-phenylene bis 5,5-dimethyl-1,3,2-dioxaborinane
23005407
2-[4-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)phenyl]-5,5-dimethyl-1,3,2-dioxaborinane
B1(OCC(CO1)(C)C)C2=CC=C(C=C2)B3OCC(CO3)(C)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 231°C to 233°C
Quantity 250 mg
Beilstein 2952607
Formula Weight 301.99
Percent Purity 99%
Chemical Name or Material 1,4-Benzenediboronic acid bis(neopentyl glycol) ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'