missing translation for 'onlineSavingsMsg'
Learn More

1-[4-(1H-Indol-3-yl)piperidino]ethan-1-one, 97%, Thermo Scientific™

Catalog Number p-2694936
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
30030-83-2
C15H18N2O
242.322
MFCD01764639
PRFIVCWVSORYRV-UHFFFAOYSA-N
1-4-1h-indol-3-yl piperidino ethan-1-one, 1-4-1h-indol-3-yl piperidin-1-yl ethanone, maybridge3_005459, 3-n-acetyl-4-piperidyl indole, 1-acetyl-4-indol-3-ylpiperidine, 3-1-acetyl-4-piperidyl indole, 1-acetyl-4-1h-indol-3-yl piperidine, 3-1-acetylpiperidin-4-yl-1h-indole, 1-4-1h-indol-3-yl-piperidin-1-yl-ethanone
2728359
1-[4-(1H-indol-3-yl)piperidin-1-yl]ethanone
CC(=O)N1CCC(CC1)C2=CNC3=CC=CC=C32
This item is not returnable. View return policy
30030-83-2
C15H18N2O
242.322
MFCD01764639
PRFIVCWVSORYRV-UHFFFAOYSA-N
1-4-1h-indol-3-yl piperidino ethan-1-one, 1-4-1h-indol-3-yl piperidin-1-yl ethanone, maybridge3_005459, 3-n-acetyl-4-piperidyl indole, 1-acetyl-4-indol-3-ylpiperidine, 3-1-acetyl-4-piperidyl indole, 1-acetyl-4-1h-indol-3-yl piperidine, 3-1-acetylpiperidin-4-yl-1h-indole, 1-4-1h-indol-3-yl-piperidin-1-yl-ethanone
2728359
1-[4-(1H-indol-3-yl)piperidin-1-yl]ethanone
CC(=O)N1CCC(CC1)C2=CNC3=CC=CC=C32
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 242.32
Percent Purity 97%
Chemical Name or Material 1-[4-(1H-Indol-3-yl)piperidino]ethan-1-one
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'