Learn More
1,3-Dioxane, 98%, Thermo Scientific Chemicals
Catalog Number p-7035883
580.82 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
505-22-6
C4H8O2
88.106
MFCD00006566
VDFVNEFVBPFDSB-UHFFFAOYSA-N
m-dioxane, 1,3-dioxacyclohexane, meta-dioxane, 1,3-propanediol formal, m-dioxin, dihydro, trimethylene glycol methylene ether, dihydro-m-dioxin, unii-b2c8m17i09, ccris 5911, 2,4-dioxane
10450
CHEBI:46924
1,3-dioxane
C1COCOC1
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
505-22-6
C4H8O2
88.106
MFCD00006566
VDFVNEFVBPFDSB-UHFFFAOYSA-N
m-dioxane, 1,3-dioxacyclohexane, meta-dioxane, 1,3-propanediol formal, m-dioxin, dihydro, trimethylene glycol methylene ether, dihydro-m-dioxin, unii-b2c8m17i09, ccris 5911, 2,4-dioxane
10450
CHEBI:46924
1,3-dioxane
C1COCOC1
1,3-Dioxane has been employed as solvent in the synthesis of 8- and 9-membered dioxazocines and dioxazonines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,3-Dioxane has been employed as solvent in the synthesis of 8- and 9-membered dioxazocines and dioxazonines.
Solubility
Fully miscible in water.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point -45°C
Density 1.032
Boiling Point 105°C to 106°C
Flash Point 5°C (41°F)
Refractive Index 1.418
Quantity 5 g
UN Number UN1165
Beilstein 102532
Solubility Information Fully miscible in water.
Formula Weight 88.11
Percent Purity 98%
Chemical Name or Material 1,3-Dioxane