missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloro-2-fluorobenzene, 98%, Thermo Scientific™

Catalog Number p-7025833
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
2268-05-5
MFCD00010307
This item is not returnable. View return policy
2268-05-5
MFCD00010307

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 36°C to 40°C
Boiling Point 168°C to 169°C
Flash Point 73°C (163°F)
Molecular Formula C8H10O4
Quantity 1 g
Beilstein 1862513
Synonym ethylene diacrylate, ethylene acrylate, ethylene glycol diacrylate, acrylic acid, ethylene ester, acrylic acid, ethylene glycol diester, 2-propenoic acid, 1,2-ethanediyl ester, acrylic acid, diester with ethylene glycol, 1,2-ethanediyl diacrylate, ethylene glycol dipropenoate, unii-42q902riye
InChI Key KUDUQBURMYMBIJ-UHFFFAOYSA-N
SMILES C=CC(=O)OCCOC(=O)C=C
IUPAC Name 2-prop-2-enoyloxyethyl prop-2-enoate
Molecular Weight (g/mol) 170.16
PubChem CID 75282
Formula Weight 165
Percent Purity 98%
Chemical Name or Material 1,3-Dichloro-2-fluorobenzene
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'