Learn More
1-(3-Chloropropyl)-1,3-dihydro-2H-benzimidazol-2-one, Mikromol
Product Code. 30461036
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Product Code. 30461036
Brand: Mikromol MM0107.100025
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-(3-Chloropropyl)-1,3-dihydro-2H-benzimidazol-2-one, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-(3-Chloropropyl)-1,3-dihydro-2H-benzimidazol-2-one
CAS 62780-89-6
Analyte or Component Names 1-(3-Chloropropyl)-1,3-dihydro-2H-benzimidazol-2-one
Shipping Condition Room Temperature
Molecular Formula C10 H11 Cl N2 O
Certifications/Compliance ISO 9001
Synonym 2H-Benzimidazol-2-one, 1-(3-chloropropyl)-1,3-dihydro-, 1-(3-Chloropropyl)-1,3-dihydro-2H-benzimidazol-2-one, 1-(3-Chloropropyl)-1,3-dihydrobenzimidazol-2-one
InChI Formula InChI=1S/C10H11ClN2O/c11-6-3-7-13-9-5-2-1-4-8(9)12-10(13)14/h1-2,4-5H,3,6-7H2,(H,12,14)
Solution Type Neat
SMILES ClCCCN1C(=O)Nc2ccccc12
IUPAC Name 3-(3-chloropropyl)-1H-benzimidazol-2-one
Molecular Weight (g/mol) 210.66
Quantity 25 mg
Formula Weight 210.056 g/mol