missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Bromobenzyl)-4-methylperhydro-1,4-diazepine, 97%, Thermo Scientific™

Catalog Number p-3522395
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
414885-80-6
C13H19BrN2
283.21
MFCD00811036
XUIRKBKKWOVRIJ-UHFFFAOYSA-N
1-3-bromobenzyl-4-methylperhydro-1,4-diazepine, 1-3-bromophenyl methyl-4-methyl-1,4-diazepane, 1-3-bromobenzyl-4-methylhomopiperazine, 1h-1,4-diazepine,1-3-bromophenyl methyl hexahydro-4-methyl, 1-3-bromobenzyl-4-methyl-1,4-diazepane, cambridge id 5266396
764720
1-[(3-bromophenyl)methyl]-4-methyl-1,4-diazepane
CN1CCCN(CC2=CC(Br)=CC=C2)CC1
This item is not returnable. View return policy
414885-80-6
C13H19BrN2
283.21
MFCD00811036
XUIRKBKKWOVRIJ-UHFFFAOYSA-N
1-3-bromobenzyl-4-methylperhydro-1,4-diazepine, 1-3-bromophenyl methyl-4-methyl-1,4-diazepane, 1-3-bromobenzyl-4-methylhomopiperazine, 1h-1,4-diazepine,1-3-bromophenyl methyl hexahydro-4-methyl, 1-3-bromobenzyl-4-methyl-1,4-diazepane, cambridge id 5266396
764720
1-[(3-bromophenyl)methyl]-4-methyl-1,4-diazepane
CN1CCCN(CC2=CC(Br)=CC=C2)CC1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 283.21
Percent Purity 97%
Chemical Name or Material 1-(3-Bromobenzyl)-4-methylperhydro-1, 4-diazepine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'