missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trimethyl-1H-pyrazole-4-carboxaldehyde, 98%, Thermo Scientific Chemicals

Catalog Number p-7033174
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
2644-93-1
C7H10N2O
138.17
MFCD00159623
HMTUBXVXHHITGO-UHFFFAOYSA-N
1,3,5-trimethyl-1h-pyrazole-4-carbaldehyde, 1,3,5-trimethyl-1h-pyrazole-4-carboxaldehyde, 1h-pyrazole-4-carboxaldehyde, 1,3,5-trimethyl, n-methyl-3,5-dimethyl-4-formylpyrazole, 1,3,5-trimethyl-1h-pyrazole-4-carbaldehyde?, akos pao-0265, timtec-bb sbb000012, acmc-1cq3n, art-chem-bb b006543, ksc496q8h
736515
1,3,5-trimethylpyrazole-4-carbaldehyde
CC1=C(C(=NN1C)C)C=O
This item is not returnable. View return policy
2644-93-1
C7H10N2O
138.17
MFCD00159623
HMTUBXVXHHITGO-UHFFFAOYSA-N
1,3,5-trimethyl-1h-pyrazole-4-carbaldehyde, 1,3,5-trimethyl-1h-pyrazole-4-carboxaldehyde, 1h-pyrazole-4-carboxaldehyde, 1,3,5-trimethyl, n-methyl-3,5-dimethyl-4-formylpyrazole, 1,3,5-trimethyl-1h-pyrazole-4-carbaldehyde?, akos pao-0265, timtec-bb sbb000012, acmc-1cq3n, art-chem-bb b006543, ksc496q8h
736515
1,3,5-trimethylpyrazole-4-carbaldehyde
CC1=C(C(=NN1C)C)C=O

1,3,5-Trimethyl-1H-pyrazole-4-carboxaldehyde is used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Trimethyl-1H-pyrazole-4-carboxaldehyde is used as organic chemical synthesis intermediate.

Notes
Air sensitive. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 80°C to 83°C
Boiling Point 132°C (12 mmHg)
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 138.17
Percent Purity 99%
Chemical Name or Material 1,3,5-Trimethyl-1H-pyrazole-4-carboxaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'