Learn More
1,2-Dimethyl-5-nitroimidazole, 97%, Thermo Scientific Chemicals
Catalog Number p-7045599
Click to view available options
Quantity:
5 g
25 g
551-92-8
C5H7N3O2
141.13
MFCD00047046
IBXPYPUJPLLOIN-UHFFFAOYSA-N
dimetridazole, 1,2-dimethyl-5-nitro-1h-imidazole, dimetridazol, emtrylvet, emtrymix, emtryl, unizole soluble, 1h-imidazole, 1,2-dimethyl-5-nitro, 5-nitro-1,2-dimethylimidazole, dimetridazolo
3090
CN1C(C)=NC=C1[N+]([O-])=O
551-92-8
C5H7N3O2
141.13
MFCD00047046
IBXPYPUJPLLOIN-UHFFFAOYSA-N
dimetridazole, 1,2-dimethyl-5-nitro-1h-imidazole, dimetridazol, emtrylvet, emtrymix, emtryl, unizole soluble, 1h-imidazole, 1,2-dimethyl-5-nitro, 5-nitro-1,2-dimethylimidazole, dimetridazolo
3090
CN1C(C)=NC=C1[N+]([O-])=O
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 136°C to 138°C
Quantity 5 g
Beilstein 130665
Merck Index 14,3265
IUPAC Name 1,2-dimethyl-5-nitroimidazole
Formula Weight 141.13
Percent Purity 97%
Chemical Name or Material 1,2-Dimethyl-5-nitroimidazole
compliance-icons compliance-icons
Product Identifier
  • 1,2-Dimethyl-5-nitroimidazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 1,2-dimethyl-5-nitro-1H-Imidazole