Learn More
1,2-Diiodotetrafluorobenzene, 99%, Thermo Scientific Chemicals
Catalog Number p-7077227
Click to view available options
Quantity:
1 g
5 g
2708-97-6
C6F4I2
401.87
JQBYIZAYQMMVTO-UHFFFAOYSA-N
1,2-diiodotetrafluorobenzene, o-diiodotetrafluorobenzene, acmc-1cuq5, 1 2-diiodotetrafluorobenzene
75923
1,2,3,4-tetrafluoro-5,6-diiodobenzene
C1(=C(C(=C(C(=C1F)I)I)F)F)F
2708-97-6
C6F4I2
401.87
JQBYIZAYQMMVTO-UHFFFAOYSA-N
1,2-diiodotetrafluorobenzene, o-diiodotetrafluorobenzene, acmc-1cuq5, 1 2-diiodotetrafluorobenzene
75923
1,2,3,4-tetrafluoro-5,6-diiodobenzene
C1(=C(C(=C(C(=C1F)I)I)F)F)F
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 49.0°C to 50.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 401.87
Percent Purity 99%
Chemical Name or Material 1,2-Diiodotetrafluorobenzene
compliance-icons
Product Identifier
  • 1,2-Diiodotetrafluorobenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.