missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloro-4-fluorobenzene, 98+%, Thermo Scientific Chemicals

Catalog Number p-7081368
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
1435-49-0
C6H3Cl2F
164.99
MFCD00018119
QSDKXMVGRLVIQV-UHFFFAOYSA-N
3,4-dichlorofluorobenzene, 3,4-dichloro-1-fluorobenzene, benzene, 1,2-dichloro-4-fluoro, pubchem3453, 3,4 dichlorofluorobenzene, acmc-1byo2, ksc494g9b, 1,2-dichloro-4-fluoro-benzene
74028
1,2-dichloro-4-fluorobenzene
FC1=CC=C(Cl)C(Cl)=C1
This item is not returnable. View return policy
1435-49-0
C6H3Cl2F
164.99
MFCD00018119
QSDKXMVGRLVIQV-UHFFFAOYSA-N
3,4-dichlorofluorobenzene, 3,4-dichloro-1-fluorobenzene, benzene, 1,2-dichloro-4-fluoro, pubchem3453, 3,4 dichlorofluorobenzene, acmc-1byo2, ksc494g9b, 1,2-dichloro-4-fluoro-benzene
74028
1,2-dichloro-4-fluorobenzene
FC1=CC=C(Cl)C(Cl)=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -1°C
Density 1.403
Boiling Point 168°C to 171°C
Flash Point 64°C (147°F)
Refractive Index 1.524
Quantity 10 g
Beilstein 1861278
Formula Weight 165
Percent Purity ≥98%
Chemical Name or Material 1,2-Dichloro-4-fluorobenzene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'