missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dibromo-3,5-difluorobenzene, 98%, Thermo Scientific™

Catalog Number p-7028362
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
139215-43-3
C6H2Br2F2
271.887
MFCD00042559
GABNJPUNFZFOJE-UHFFFAOYSA-N
benzene, 1,2-dibromo-3,5-difluoro, 1,2-dibromo-3,5-difluoro-benzene, pubchem3438, acmc-1c3vh, 2,3-dibromo-1,5-difluorobenzene, 1,2-bis bromanyl-3,5-bis fluoranyl benzene
2724517
1,2-dibromo-3,5-difluorobenzene
C1=C(C=C(C(=C1F)Br)Br)F
This item is not returnable. View return policy
139215-43-3
C6H2Br2F2
271.887
MFCD00042559
GABNJPUNFZFOJE-UHFFFAOYSA-N
benzene, 1,2-dibromo-3,5-difluoro, 1,2-dibromo-3,5-difluoro-benzene, pubchem3438, acmc-1c3vh, 2,3-dibromo-1,5-difluorobenzene, 1,2-bis bromanyl-3,5-bis fluoranyl benzene
2724517
1,2-dibromo-3,5-difluorobenzene
C1=C(C=C(C(=C1F)Br)Br)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 36°C to 38°C
Density 2.13
Flash Point >110°C (230°F)
Quantity 5 g
Formula Weight 271.89
Percent Purity 98%
Chemical Name or Material 1,2-Dibromo-3,5-difluorobenzene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'