missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Chloronicotinoyl)piperidine, 97%, Thermo Scientific™

Catalog Number p-7033689
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
56149-33-8
C11H13ClN2O
224.688
MFCD03387556
PHTPEDJMJRDQMY-UHFFFAOYSA-N
2-chloro-3-piperidin-1-ylcarbonyl pyridine, 1-2-chloronicotinoyl pyridine, 2-chloropyridin-3-yl piperidin-1-yl methanone, 2-chloro-3-piperidine-1-carbonyl pyridine, 2-chloro-pyridin-3-yl-piperidin-1-yl-methanone, 2-chloro 3-pyridyl piperidyl ketone, acmc-20ao0i, 1-2-chloronicotinoyl piperidine, 2-chloro-3-1-piperidinylcarbonyl pyridine, 2-chloro-3-piperidin-1-yl carbonyl pyridine
4584285
(2-chloropyridin-3-yl)-piperidin-1-ylmethanone
C1CCN(CC1)C(=O)C2=C(N=CC=C2)Cl
This item is not returnable. View return policy
56149-33-8
C11H13ClN2O
224.688
MFCD03387556
PHTPEDJMJRDQMY-UHFFFAOYSA-N
2-chloro-3-piperidin-1-ylcarbonyl pyridine, 1-2-chloronicotinoyl pyridine, 2-chloropyridin-3-yl piperidin-1-yl methanone, 2-chloro-3-piperidine-1-carbonyl pyridine, 2-chloro-pyridin-3-yl-piperidin-1-yl-methanone, 2-chloro 3-pyridyl piperidyl ketone, acmc-20ao0i, 1-2-chloronicotinoyl piperidine, 2-chloro-3-1-piperidinylcarbonyl pyridine, 2-chloro-3-piperidin-1-yl carbonyl pyridine
4584285
(2-chloropyridin-3-yl)-piperidin-1-ylmethanone
C1CCN(CC1)C(=O)C2=C(N=CC=C2)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 61°C to 62°C
Quantity 250 mg
Formula Weight 224.69
Percent Purity 97%
Chemical Name or Material 1-(2-Chloronicotinoyl)piperidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'