missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Thiadiazole-4-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-7025394
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
4100-13-4
C3H2N2O2S
130.121
MFCD00052099
HJZYBDPHAHGHAZ-UHFFFAOYSA-N
1,2,3-thiadiazole-4-carboxylic acid, 1,2,3 thiadiazole-4-carboxylic acid, thiadiazolecarboxylicacid, 4-carboxy-1,2,3-thiadiazole, 1,2,3-thiadiazole4-carboxylic acid, 1 pound not2 pound not3 thiadiazole-4-carboxylicacid
351418
thiadiazole-4-carboxylic acid
C1=C(N=NS1)C(=O)O
This item is not returnable. View return policy
4100-13-4
C3H2N2O2S
130.121
MFCD00052099
HJZYBDPHAHGHAZ-UHFFFAOYSA-N
1,2,3-thiadiazole-4-carboxylic acid, 1,2,3 thiadiazole-4-carboxylic acid, thiadiazolecarboxylicacid, 4-carboxy-1,2,3-thiadiazole, 1,2,3-thiadiazole4-carboxylic acid, 1 pound not2 pound not3 thiadiazole-4-carboxylicacid
351418
thiadiazole-4-carboxylic acid
C1=C(N=NS1)C(=O)O

It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents, reducing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Melting Point 227°C to 228°C
Quantity 1 g
Beilstein 113707
Solubility Information Slightly soluble in water.
Formula Weight 130.12
Percent Purity 97%
Chemical Name or Material 1,2,3-Thiadiazole-4-carboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'